Would you like to log in again?
Hermle Universal Benchtop Centrifuges
Supplier: Hermle
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Z326-0685-25 Z326-1050 Z326-1205 Z326-6405 Z326-2010 Z326-1215 Z306-100 Z326-02MP Z326-3015 Z326-3020H Z326-4420 Z326-100H Z326-2420H Z326-3202 Z326-2420 Z326-0650-HS Z326-K-BND Z326-0650-LS Z326 Z326-K Z326-100H-AC Z326-BND
470230-648EA 1837 USD InStock
470230-648 470230-658 470230-668 470230-646 470230-678 470230-666 470230-644 470230-654 470230-676 470336-498 470230-664 470230-674 470230-652 470230-684 470230-662 470230-672 470230-650 470230-682 470230-660 470230-670 470230-680 470336-500
Hermle Universal Benchtop Centrifuges
Centrifuges Benchtop Centrifuges
Setting new standards in this centrifuge class.
  • Bundles are available with our most popular rotor choices
  • Models available with or without refrigeration
  • 19 different rotor choices available to choose from
  • Suitable for nearly every laboratory application
With 19 available rotor options, the Z326 defines the term “universal”. Swinging bucket rotors and high speed angled rotors are available to accept tubes from 0.2 ml up to 85 ml, making this centrifuge suitable for nearly any laboratory application. Despite an impressive capacity of up to 4×100 ml (or 6×85 ml), the unit is surprisingly compact with both the standard air-cooled model and refrigerated model featuring a width of just over 15".
Advanced microprocessor controls allow for instant rotor identification, which automatically converts the rotational speed to the relevant g-force (rcf), preventing the user from setting a speed above the capabilities of the installed rotor. In addition to retrieving saved programs, the easy to use front panel can adjust accel/decel rates, and change the end of run alarm tone.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty