Would you like to log in again?
Bradley Pressurized Portable Eye Wash
Ward's
AA585
470231-110EA 3530.7 USD InStock
470231-110
Bradley Pressurized Portable Eye Wash
First Aid Supplies Eyewash Stations and Eyewash Bottles
Easily transportable for fast, efficient treatment.
  • Built-in handles allow for easy transportation
  • Includes dual eyewash heads for flow control and pop-off dust covers
  • Eyewash has supplemental 8" yellow hose with hand-held squeeze valve to spray face or rinse body
  • Stainless steel tanks feature pressure relief valve and tire filter valve with air gauge
  • Refill heater jackets provide freeze protection down to –20 °F (–29 °C)
These eye washes are ideal for providing emergency protection in areas where permanent water supply is not readily available or accessible. Supplements permanent eyewash fixture installations. Ten-gallon pressure tank is constructed of stainless steel. Eyewash heads are chrome-plated brass. Tire filler valve with 0-160 PSI air gauge. 8" hand-held hose spray has chrome-plated metal parts. Hose burst strength is 450 PSI. Hand-held hose spray head contains an antimicrobial agent to protect the spray head.
Certifications: Meets ANSI Z358.1-2009 Standards, SEI Certified.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty