Would you like to log in again?
Ocean Trash Can
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
Ward's®
9-120
470231-166EA 125.5 USD InStock
470231-166
Ocean Trash Can
Educational Materials Environmental Educational Materials Environmental Learning Activities
This exercise examines various types of pollutants, their sources, and their effects on the ocean, its inhabitants and the surrounding coastline.
  • Grades 7-10
  • Designed for 24 students working in groups of 4
  • Requires 4 class periods
Students classify ocean pollution into five basic categories; debris, toxins, dissolved nutrients, particulates, and oil. They model the effects of ocean pollution with activities examining a variety of topics: the effect of the pH of various liquids on calcium carbonate; the effects of dilution, buffering capacity, salt and acid runoff; acid rain; the ability of water to purify itself; and oil spill clean-up. As a final project, students design and write a newsletter containing articles with pertinent pollution topics such as current laws, pollution, effects, bioaccumulation, and prevention.
Ordering information: Six 1 gallon milk containers are required, but not included. Kit contains one instructor Manual, Student Data Sheets, Plastic Cups, Cupcake Liners, Sand, Tums, pH Strips, Radish Seeds, Sea Salt, Styrofoam Peanuts, Feathers, Yarn, Plastic Bags, Cotton Balls, Animal Fur, Wire.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty