Would you like to log in again?
Neulog Bluetooth Connection Device with Built-in Battery
Supplier: EISCO SCIENTIFIC LLC
BLT202
470231-416EA 136.75 USD InStock
470231-416
Neulog Bluetooth Connection Device with Built-in Battery
Data Loggers
The Neulog Bluetooth connection device lets you connect any Neulog sensor to a iPhone, Droid, PC, Mac, or Chromebook Device using Bluetooth connectivity.
  • Allows connection of 1-5 sensors to a tablet at one time
  • Fast, easy, seamless connection of Neulog sensors to your devices
Full app for connection can be found in the Apple App Store and Google Play Store.
Neulog software allows real time streaming of data, setting experiments and collecting data over a period of time, and analyzing data with a powerful analytical package.
Caution: For educational use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty