Would you like to log in again?
Basic Inorganic and Organic Chemistry Molecular Model Set, 78 Pieces
Supplier: EISCO SCIENTIFIC LLC
CH0609
470231-422EA 21.75 USD InStock
470231-422
Basic Inorganic and Organic Chemistry Molecular Model Set, 78 Pieces
Models Molecular Biology Models
Standard Size Molecular Model Set
  • 54 atoms
  • 24 links
Contents: 24 White Hydrogen (1 Hole, 15 mm), 12 Black Carbon (4 Hole, 22 mm), 6 Red Oxygen (2 Hole, 20 mm), 2 Blue Nitrogen (4 Hole, 22 mm), 4 Blue Nitrogen (3 Hole, 22 mm), 1 Yellow Sulfur (4 Hole, 22 mm), 1 Yellow Sulfur (2 Hole, 22 mm), 2 Green Chlorine (1 Hole, 22 mm), 1 Brown Bromine (1 Hole, 22 mm), 1 Purple Iodine (1 Hole, 22 mm), 16 Medium Grey Bonds, 8 Long Grey Bonds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty