Would you like to log in again?
Adam-X
Supplier: AMERICAN 3B SCIENTIFIC SE
3010320 1021109
470231-474EA 74500.09 USD InStock
470231-474 470305-058
Adam-X
Models Human Biology Models
High Fidelity Simulator At A Great Price.
  • Ideal For trauma and emergency training
  • Wireless connectivity for monitoring a response program
  • Programmable reactions to examination and treatment
ADAM-X is the male patient simulator designed for practicing skills and providing medical assistance in case of emergency. It is human-sized, and has a very realistic skin. ADAM-X has automatic blinking depending on physiological status, as well as a physiological reaction of the pupils to light and to touch. The ADAM-X has a bilaterally palpable pulse on the carotid artery, the femoral artery, the radial artery, the dorsal artery of the foot, the popliteal artery and the posterior tibialis.
ADAM-X has a wide variety of programmable sounds. Auscultation is possible with 8 points of breath sounds, 5 points of heart sounds and 14 points of lung sounds on the front of the mannequin, with the possibility of an independent set of options of the sounds in 4 areas. There is also the possibility of auscultation on the back of the mannequin. Breath sounds are independent and are related to ventilation of each lung. ADAM-X has speech, and can say “yes”, “no” and some interjections. Also featured are guttural and coughing sounds.
ADAM-X has a very realistic respiratory tract. It has fully independent right and left lungs, and ventilation of the lung automatically leads to the appropriate sounds of breathing, chest excursion and pulmonary gas exchange. The occurrence of apnea or hypoventilation, along with the low fraction of oxygen in the inspired gas mixture, lead to corresponding physiological changes: the hypercapnia, the hypoxemia, the decrease of the SpO2 and the tachycardia. Ventilation of the lungs at a positive pressure or spontaneous ventilation restoration returns ADAM-X to the breath with an appropriate amount of respiratory rate. ADAM-X also features chest flotation, and the resistance of the airway changes accordingly
The ADAM-X is ideal for ALS training, scenario training and medical treatment training. Because the ADAM-X is completely wireless and tetherless, and has realistic human proportions, it is very portable and ideal for on-site training. During CPR training, the accuracy of the hand placement is tracked, as well as the depth and rate of compression.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty