Would you like to log in again?
Forensic Toxicology
Supplier: ALDON CORP SE
Ward's
IS9022
470231-536KT 161.7 USD InStock
470231-536
Forensic Toxicology
Educational Materials Forensics Educational Materials Trace Evidence Learning Activities
Be the Forensic Scientists.
  • Kit Includes enough materials for 30 students
  • Analyze 15 different patient samples
Your class with utilize different lab techniques to determine the presence of alcohol, illicit drugs and pain medications present in the simulated urine samples of suspected criminals. The various tests include the Marquis Test for Morphine, Simon's test for MDMA, THC Immunoassay, Cocaine Immunoassay, and 6-Monoacetylmorphone Test.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty