Would you like to log in again?
Innovating Science® Formation of a Native Copper Mineral Deposit
Supplier: ALDON CORP SE
Innovating Science®
AD-KK7105-KIT IS8701-SGL
470231-554KT 37.25 USD InStock
470231-554 470225-444
Innovating Science® Formation of a Native Copper Mineral Deposit
Educational Classroom Kits and Activities
Have you ever wondered how Copper is Formed in Nature?
  • Multi-lab day that shows the gradual growth of mineral copper deposits
  • Aligned to NGSS Standards PS1.A, PS1.B and ESS3.A
  • Contains a teacher's guide and student study and analysis copymasters
Of all the naturally occurring metals copper is one of the most useful and versatile. Evidence suggests copper has been used by humans for at least 10,000 years. Copper is used for architecture. It is used for electrical wiring, water pipes, and plumbing fixtures. It is found in automobiles and a variety of electronics and electronic devices. It is in electrical motors, generators, and turbines.
It is an important part of heat exchangers found in refrigeration and air conditioning units, and it is in circuit boards and electromagnetics. While copper is typically extracted from copper-containing mineral ores that must be treated and refined to obtain the pure metal, copper also exists in nature in its native elemental form.
This is the rarest and most valuable form of copper as it is the easiest to process to obtain pure metal. In this activity, students will create specific environmental conditions and examine the formation of native copper deposits over several days.
Delivery information: Instructor's Manual and SDS included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty