Would you like to log in again?
Diffusion and Osmosis - Small Group Learning Kit
Supplier: Aldon
IS3704-SGL
470231-558KT 95.7 USD InStock
470231-558
Diffusion and Osmosis - Small Group Learning Kit
Educational Materials Biology Educational Materials Cell Biology Learning Activities
Observe osmosis in Living Tissue.
  • Small group learning kit
  • Student copymasters and teacher guide included
Students will study the movement of water and nutrients across a cell membrane and observe osmosis in living tissue. They will then investigate the relationship between surface area and volume as it relates to cells and diffusion. They will also examine the concept of molarity and how it relates to osmotic potential and the movement of water. Students will also be able to explain how cell size and shape affect the overall rate of nutrient intake and water elimination. Students will also use plant tissue to determine the molarities of unknown solutions based on the direction and degree of water movement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty