Would you like to log in again?
Innovating Science® Mendelian Genetics: Genes and Probability Lab Activity
Supplier: ALDON CORP SE
Innovating Science®
IS3010 IS3010-SGL
470231-566KT 33.25 USD InStock
470231-566 470343-484
Innovating Science® Mendelian Genetics: Genes and Probability Lab Activity
Educational Classroom Kits and Activities
Learn about Mendelian genetics and determine patterns of inheritance.
  • Students will simulate Mendel's work and simulate both monohybrid and dihybrid crosses as well as determine genotypic and phenotypic ratios
  • Aligned to NGSS Practices
  • Performance expectations: HS-LS4-3, HS-LS4-4, HS-LS4-5
  • Disciplinary core ideas: LS4.B, LS4.C
  • Cross cutting concepts include patterns, cause and effect
  • Engineering practices include analyzing and interpreting data, constructing explanations and designing solutions, engaging in argument from evidence
In this activity, students can simulate Mendel's work and determine patterns of inheritance. Using special chips and Innovating Science's exclusive 'double dice', students will be able to simulate both monohybrid and dihybrid crosses.
After the crosses, students will be able determine genotypic and phenotypic ratios for select traits and compare their values to the theoretical 'ideal' values as put forth by Mendel. Kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery information: Instructor's Manual and SDS included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty