Would you like to log in again?
Properties of Toothpaste - Small Group Learning Kit
Supplier: ALDON CORP SE
IS9350-SGL
470231-570KT 64.35 USD InStock
470231-570
Properties of Toothpaste - Small Group Learning Kit
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
Create your own Toothpaste.
  • Small group learning kit
  • Student copymasters and teacher guide included
The history of teeth cleaning agents actually goes back thousands of years. Toothpaste as we know it today however is a comparatively recent development. In this activity, students will investigate several properties of toothpaste samples, provided in the kit, including presence of fluoride, pH, abrasiveness, and foaming ability. As a second activity, students will use the included materials to create their own toothpaste.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty