Would you like to log in again?
Science In the Kitchen - Small Group Learning Kit
Supplier: ALDON CORP SE
IS9355-SGL
470231-576KT 122.3 USD InStock
470231-576
Science In the Kitchen - Small Group Learning Kit
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
Explore chemistry that happens in the Kitchen!
  • Small group learning kit
  • Student copymasters and teacher guide included
This comprehensive kit incorporates a variety of scientific techniques all themed around common kitchen materials. Students will perform chromatography on commercial food colors, use chemical tests to identify an unknown cooking ingredient, examine the protein digesting ability of a common meat tenderizing enzyme, use titration to quantify vitamin C levels and then test an unknown juice or soda (not provided), and lastly examine some of the differences and similarities in the materials used to clean up (soap, hand dishwashing detergent, and machine dishwashing detergent).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty