Would you like to log in again?
Innovating Science® A Safer Flame Test
Supplier: ALDON CORP SE
Innovating Science®
IS1515 IS2520-SGL AD-KK7106-KIT
470225-446KT 115.15 USD InStock
470225-446 470231-582 470330-520
Innovating Science® A Safer Flame Test
Educational Materials Chemistry Educational Materials Ionic Properties Learning Activities
A metal identification lab.
  • Hands-on activity that allows students to identify different metal ions safely
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-1,2,5; HS-PS1-1,2,7
  • Disciplinary core ideas: PS1.A, PS1,B
  • Crosscutting concepts: Patterns; scale, proportion, and quantity; energy and matter
  • Engineering Practices: Developing and using models, analyzing and interpreting data
The flame test is an analytical technique often used for the identification of certain elements, primarily metal ions. In this activity, a safer method is employed, using special mineral-salt containing candles that significantly reduce the amount of alcohol required to perform the flame test experiment. Students will identify specific metal ions by comparing the emission spectra for each metal to a set of known spectra and then, based on their findings, identify a metal ion in an unknown sample. As part of this activity students will correctly identify the element emitting the colored flame by its spectra, qualitatively identify possible metals from the color of the flame being emitted, explain the process that caused the flame to change color and understand the definition of a chemical reaction.
Delivery information: Instructor's Manual and SDS included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty