Would you like to log in again?
iworx® Physiology Recording Equipment
Supplier: IWORX SYSTEMS INC.
iworx®
PTS-ANIMAL BP-100 PTS-PSYCH PTS-BASE FT-302 PTS-GENBIO PTSMETABOLICNG PTSMETABOLIC
470231-634EA 6850.2 USD InStock
470231-634 470231-636 470231-658 470231-640 470231-642 470231-660 470231-682 470231-638
iworx® Physiology Recording Equipment
Models Human Biology Models
Easy To Use Physiological Recording.
  • Includes Labscribe software and activities
  • Expand the basic kit with additional probe bundles
The iworx® physiological recording system is high quality and easy to use. The base kit allows you to perform more than 40 experiments, and 100 exercises with the expertly written activates contained in the LabScribe software. Each activity walks the students through equipment setup, probe attachment and data collection. The base unit can then be expanded using additional sensors in the expansion kits. Each Expansion kit extends the amount of experiments and exercises you can perform in class.
The Base Kit includes the control module with built-in Spirometer, heart sounds sensor, Grip force/Blood Pressure sensor, Temperature Sensor, LabScribe software, ECG /EMG recording module, Hand Dynamometer, Event Marker, Pulse Plethysmograph, temperature probe, blood pressure cuff, stimulator cable, recording leads, and disposable electrodes. These basic tools allow you to perform over 40 activities included in the software package, on topics like Human circulation, spirometry, The human nerve; Exercise, the heart, and psychophysiology.
The number of activities can be expanded using the expansion kits. The animal kit gives you 9 more probes and cables of studying small animal physiology and allowing you to use 20 more prebuilt activities. The Psychology bundle gives you 5 more probes, 13 more activities, and a chance to show students how physiology reacts to psychological stress. The exercise physiology set is available with or without calibration gases, and allows you to measure gas differentials during exercise. Finally the General Bio set gives you 3 more probes and a respiration chamber, allowing for generalized recordings of plant and animal specimens.
PC requirements:
LabScribe Software System Requirements: Windows: Vista, 7, 7, 10; Mac: OSX,10.9, above;
Min Requirements: Pentium IV 1GHz, 2GB RAM, 2GB Free Hard drive Space, 2 Available USB ports;
Recommended Specs: Quad Core 64 bit processor, 4GB or More Memory
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty