Would you like to log in again?
iworx® Eight Channel Data Recorder
Supplier: IWORX SYSTEMS INC.
IX-RA-834
470231-644EA 6137.3 USD InStock
470231-644
iworx® Eight Channel Data Recorder
Data Loggers
The iWorx RA Recorder is the most advanced and feature-rich physiological data acquisition system on the market. It comes standard with eight general purpose input channels, a low voltage and high voltage stimulator, eight digital inputs and outputs, a built-in barometric pressure sensor, and four iWire™ inputs.
  • Flexible recording
  • 16 bit resolution
  • 4 BNC inputs, 4 DIN8, and 4 wire inputs
  • High and low voltage stimulator output
The iWorx RA exhibits the high resolution, low noise, and flexibility required for a variety of research applications including cardiac function, themodynamics, isolated tissue and organ studies, exercise physiology research, metabolic function studies, neuroscience and both oocyte and epithelial voltage clamp applications. Incorporating innovative iWire serial interface technology and advanced LabScribe data acquisition and analysis software, the iWorx RA Recorder accommodates a wide range of sensors, transducers, and other devices.
LabScribe Software System Requirements: Windows: Vista, 7, 7, 10; Mac: OSX,10.9, above;
Min. Requirements: Pentium IV 1GHz, 2GB RAM, 2GB Free Hard drive Space, 2 Available USB ports;Recommended Specs: Quad Core 64 bit processor, 4GB or More Memory
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty