Would you like to log in again?
iworx® Small Animal ECG / EMG Systems
Supplier: IWORX SYSTEMS INC.
IX-BIO4-SA IX-BIO8-SA
470231-656EA 8408.15 USD InStock
470231-656 470231-654
iworx® Small Animal ECG / EMG Systems
Data Loggers
This system records small animal data easily. A four or eight channel biopotential amplifier that allows multiple channels of EMG, ECG, EOG to be recorded simultaneously from a single subject.
  • 4 or 8 channel recording
  • USB power and data connection
  • Includes Labscribe software
Each recorder is furnished with LabScribe Recording and Analysis Software and connects directly to a USB port on either a Macintosh or Windows PC. Measuring 2 channels of ECG, Lead I and Lead II allows LabScribe to calculate Lead III, aVR, aVF and aVL. EMG signals can be measured at the same time as the ECG signals.
LabScribe Software System Requirements: Windows: Vista, 7, 7, 10; Mac: OSX,10.9, above;
Min. Requirements: Pentium IV 1GHz, 2GB RAM, 2GB Free Hard drive Space, 2 Available USB ports; Recommended Specs: Quad Core 64 bit processor, 4GB or More Memory
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty