Would you like to log in again?
INVERTED MICROSCOPE TRI INFINITY OPTICS
Supplier: LW SCIENTIFIC SE
Inverted Microscope trinocular^ Inverted Microscope, trinocular, infinity optics, 4x/40x plan objectives, 10x/20x plan phase objectives, halogen illumination, C-mount video adapter, Mechanical stage, Power Supply: 90/240VAC, 50/60Hz, Includes: Dust Cover, Objectives, Abbe Condenser, Table: 160x250mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INVERTED MICROSCOPE TRI INFINITY OPTICS 470231-944 L1M-T04A-DPH3 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty