Would you like to log in again?
Minerals Model Set
Supplier: Mega Molecules
GC430
470232-074EA 61.5 USD InStock
470232-074
Minerals Model Set
Models Crystal Models
Hands on mineral crystal structures.
  • Includes 1 activity
  • Perfect for high school and college students
Build the most abundant silicate minerals including feldspar, quartz, olivine, pyroxenes, amphiboles, micas, and clay minerals. Learn how models of silicate minerals are related to unit cell and crystal lattice structures. This model set has enough parts to construct all six types of silicate minerals at one time allowing students to compare nesosilicates, sorosilicates, cyclosilicates, inosilicates, phyllosilicates, and tectosilicates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty