Would you like to log in again?
Neulog Sense Infrared Transmitter
Supplier: EISCO SCIENTIFIC LLC
SNS160
470232-636EA 51.3 USD InStock
470232-636
Neulog Sense Infrared Transmitter
Educational Materials Physics Educational Materials Applied Physics Learning Activities
An accessory for your SENSE bot!
  • Neulog sense infrared transmitting unit
  • Great for sending an IR sensor to use in group robotic events, such as building Sense bot traffic, or battle bots
  • Easy USB connection to Neulog Sense
  • Recommended for use with the Neulog Sense Tracking Unit which can be purchased separately
The Neulog Sense infrared transmitter is the sister to the tracking unit. The transmitter outputs an IR light that the Neulog Sense infrared tracking unit can locate. It is great for coding robots to follow IR signals or build follow and chase scenarios with multiple sense bots. Most commonly used in an autonomous car challenge where one Sense bot needs to follow another, much like adaptive cruise control in many of todays modern cars.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty