Would you like to log in again?
Neulog Sense Servo Gripper Arm
Supplier: EISCO SCIENTIFIC LLC
SNS167
470232-638EA 122.59 USD InStock
470232-638
Neulog Sense Servo Gripper Arm
Educational Materials Physics Educational Materials Applied Physics Learning Activities
An accessory for your SENSE bot!
  • Gripper turns sense into a robot that can modify its environment
  • The gripper has 2 servomotors, one to move the gripper up and down, the other to open and close
  • Each servo of the gripper has a transmission gear and potentiometer
  • Requires a Neulog SENSE to run
  • Gripper received various commands you can program: up-mid-down, open, close
Allow your Sense to monitor and change its environment with the Sense Gripper. The gripper has two servomotors, one to move the gripper up and down, the other to open and close. Each servo of the gripper has a transmission gear and potentiometer. With block programming it is easy to operate the gripper with commands within the block code. An example of these commands are Up-Mid-Down, Open-Close. Be creative and activate the gripper using other Neulog sensors you can plug into sense, for example use the sound sensor to activate the gripper with a clap.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty