Would you like to log in again?
Neulog Sense C Language Coding Unit
Supplier: EISCO SCIENTIFIC LLC
CARM202
470232-640EA 126.39 USD InStock
470232-640
Neulog Sense C Language Coding Unit
Educational Materials Physics Educational Materials Applied Physics Learning Activities
The Sense C is a trainer that allows a user to study and exercise C language and to program Neulog Sense in C language.
  • Sense is a STEM computer programing and engineering learning platform
A student from the age of 8 years old can begin programming sense via visual block programming (free for download) and then solve any challenge imaginable by plugging various sensors, coding modules, or robotic parts into sense.
Once a student grasps the fundamentals of visual programing they can then purchase modules to program in a variety of other languages such as C and Python. Sense will provide STEM and coding education from ages 8 – 21.
C is the coding language for creating machine programs and is the most needed language in the industry right now. Students can use the Sense C language programming unit to train on how to code in C an then program there Sense bots and Neulog sensors to solve real world STEM problems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty