Would you like to log in again?
Wallcur® PRACTI-Insulin
Supplier: Wallcur
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
47030 445NL 409HM 405TP 410LT SP-419 414VP 452HMG 408VP 9405REG 9405NPH
470232-894BX 88.99 USD InStock
470232-894 470232-882 470233-104 470232-810 470232-820 470233-106 470232-698 470232-804 470232-814 470232-812 470233-230
Wallcur® PRACTI-Insulin
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Practice With A Wide Variety Of Insulin Products
  • Contains No Medication
  • Multiple Options Available
Teaching insulin injections starts with the fundamentals of carefully reading the insulin vial labels for dosage information as many insulin labels look alike. With PRACTI- Insulin you can teach your students how to calculate dosages, choose the applicable syringe for the dosage, draw up the correct amount of air, and then rotate, mix and withdraw before the medication precipitates out. Wallcur’s PRACTI- Insulin Vials contain distilled water and an additive that closely simulates the real thing and must be mixed properly, and administered promptly.
Teach your students to:
Identify and understand two of the most commonly ordered insulin medications
Read and interpret differing insulin labels that often look-a-like
Calculate insulin dosages
Mix, combine, and promptly withdraw short and long acting insulin
Identify injection site landmarks, and aseptic precautions when administering insulin
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty