Would you like to log in again?
Wallcur® PRACTI-Patches
Supplier: Wallcur
8100DF
470233-072PK 98.79 USD InStock
470233-072
Wallcur® PRACTI-Patches
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Teach Important Skills Of Medication Application
  • Contains No Medication
  • Available With 2 Different Labels For Training
The Wallcur PRACTI-Patches teaches your students the critical skills of transdermal medication infusion. We have carefully manufactured and labeled each type of Patch to closely resemble the "real thing" simulating the generic drug Fentanyl (Duragesic®) or Nitroglycerine (Nitro-Stat® and Nitro-Dur®). This training provides the most authentic simulation practice interpreting and converting between systems of measure, calculating normal drug dosages, and determining the infusion dosage per hour.
Hands on application and removal of the PRACTI-Patches gives students the opportunity to practice without exposure to the real medication.
An excellent lab skill practice for students of nursing, pharmacy tech, medical assisting, EMT and more.
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty