Would you like to log in again?
Somso® Nursing Care Manikins
Supplier: Somso
CLA2 CLA1
470233-236EA 5292.2 USD InStock
470233-236 470233-238
Somso® Nursing Care Manikins
Models Human Biology Models
For more advanced patient care practice!
  • A great starter manikin
  • Expandable over time
Standard nursing doll in natural size for nursing care comprising of the following parts: Removable head, removable eyes, dentures and tracheostomy, thorax with chest wall and moving arms with infusion and injection pads, lower body with abdominal wall and anus praeter abdominal wall, injection pads in the buttocks, movable mounted legs with injection pads in the thigh, lung alveolus, stomach, intestines, detachable female and male genitalia with rectum and bladder.
The advanced version includes lungs exhibiting closure, additional male genetalia and a higher capacity bladder (2 L).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty