Would you like to log in again?
Somso® Intubation Manikin
Supplier: Somso
CLA8
470233-264EA 5205.75 USD InStock
470233-264
Somso® Intubation Manikin
Models Human Biology Models
An intubation model with feedback!
  • Can be combined with the CLA Nursing Doll
  • Provides alarm if to much pressure is exerted
  • No mechanics makes confirmation of success a stethoscope away
Natural size. This functional model, which has been developed in cooperation with the Federal Centre for Health Education in Cologne, makes it possible to learn how to intubate under life-like conditions. In its design and construction special importance has been placed on natural size and anatomical features to develop a functional model. If during intubation too much pressure is applied through the laryngoscope to the upper incisors, an acoustic signal is heard. The correct location of the tube, inserted after intubation, may be confirmed by audible respiratory sounds through a stethoscope lateral to the left and right wall of the thorax.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty