Would you like to log in again?
Somso® Newborn Intubation Manikin
Supplier: Somso
CLA8/58
470233-266EA 2213.8 USD InStock
470233-266
Somso® Newborn Intubation Manikin
Models Human Biology Models
Easy to use!
  • Simple assembly
  • Naso or Orotracheal intubation practice
With the co-operation of Prof. Dr. Chr. Fusch, Uniklinik Greifswald, we have developed a life-size model for orotracheal and nasotracheal intubation. In addition to a moveable lower jaw, great importance has been placed on faithfully copying the nasal, oral and pharyngeal cavities.
To practice nasotracheal intubation, a tube is carefully introduced into the nostril and carefully pushed along the inferior meatus and the back wall of the throat to the 7 cm mark, to ensure that the point of the tube stops shortly before the epiglottis. Without any pressure being exerted on the upper jaw, the base of the tongue is gently raised using a laryngoscope spatula held in the left hand, until the entrance to the oesophagus and the glottis is visible. Holding the Magill forceps in the right hand, the point of the tube is then taken and inserted approximately 1.5 cm into the trachea via the glottis.
We recommend that to prevent the tube from accidentally slipping, the thumb and index finger of the left hand hold the tube at the nasal vestibule, and the remaining fingers and palm of the hand are placed on the left side of the temple and forehead of the new-born baby. The tube can then be attached to the skin as usual (e.g. using adhesive tape). The tube can be introduced through the mouth directly into the trachea, when the trachea is visible for orotracheal intubation.
Components:
- Movable head with lifelike nasal cavity, mouth and throat, movable tongue and jaw with lifelike trunk with diaphragm and soft movable arms and legs
- Pulmonary alveoli filled with foam material
- Stomach
- Female thoracic and abdominal cover with umbilical cord
- Pressure spray bottle with silicone oil (125 mL)
- Instructions for use
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty