Would you like to log in again?
Somso® Broncho-Junior Trainer
Supplier: Somso
CLA9/23
470233-276EA 4168 USD InStock
470233-276
Somso® Broncho-Junior Trainer
Models Human Biology Models
Pediatric bronchoscopy practice.
  • Simulates a 3 – 5 year old child
  • Can also be used for intubation practice
Life-sized, in plastic. Intubation trainers for pediatric and adult bronchoscopy, ICU and anesthesiology. The Broncho Boy family of mannequins presents specific solutions for learning and improving intubation skills in a wide range of hands-on teaching situations. The most recent addition to the group is Broncho Junior, a pediatric trainer with physical characteristics of a three-to five-yearold child.
Broncho Junior was designed and constructed with the following teaching goals in mind:
• Facilitate Introduction to flexible and rigid pediatric bronchoscopy
• Provide help in learning more advanced procedures, such as foreign body extraction and stent placement
• Help pediatric anesthesiology and ICU trainees to master naso- and oro-tracheal intubation while using the flexible bronchoscope as a guide
Components :
1. Head with ribcage
2. Movable fitted head with nasal cavity, mouth and throat
3. Larynx with removable bronchial tree
4. Chest cover
5. Sternum with auscultation membranes on right and left 6. Trunk with folding hinge mounted on baseboard
7. Silicone oil, 125 mL pressure spray bottle
8. Instructions for use
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty