Would you like to log in again?
Somso® Nursing Child Manikins
Supplier: Somso
CLA23/A CLA23/B CLA23
470233-298EA 3856.7 USD InStock
470233-298 470233-300 470233-302
Somso® Nursing Child Manikins
Models Human Biology Models
Easy to use 3 year old Manikin.
  • Interchangable parts
  • Moveable limbs
  • Available in 3 versions
Natural size. The doll with a length of 90 cm. and weighing 6 kg corresponds to a small child approximately 3 years old. This realistic training doll offers the facility for numerous nursing procedures for training paediatric nurses.
This doll, made of plastic, incorporates durable, new style joint mechanisms which permit most natural movements. As a result training and nursing procedures can be carried out in most practical situations.
This small child phantom is available as both a female and male model because of the interchangeable chest and abdominal wall. The abdominal walls incorporate application points for S.C. injections and treatment of wounds (female abdominal wall - OP wound after appendectomy, male abdominal wall - wound treatment after hernia). Upper arms and upper thighs are provided with injection pads. The right arm can be exchanged for an I.V. injection arm.
The abdominal walls are equipped with:
- anus-praeter
- openings for PEG and suprapubic bladder puncture.
The CLA Child Nursing Doll offers the following possibilities for training and instruction:
General
Nursing procedures
a) Dressing and undressing
b) Daily body care
c) Movement, putting to bed and positioning
d) Mouth and teeth care
e) Eye, nose and throat care (including administration of drops)
I. Enema
a) Lavage enema / clyster
b) Intestine tube insertion
II. Catheterising and feeding
a) Demonstration of catherisation and permanent catheter insertion in female and male genitalia
b) Stomach tube insertion through nose and mouth
c) Nutrition through bolus administration or by nutrition pump
III. Injections and infusions
a) I.M. injection (thigh, upper arm)
b) S.C. injection (stomach)
c) Demonstration of I.V. injection and infusion
d) Connection of infusion equipment
e) I.V. puncture
IV. Lavage
a) Gastric lavage
V. Wound treatment, catheter care and other measures
a) Wound care and suture removal
b) PEG care
c) Anus-praeter care
d) Suprapubic bladder puncture with material removal and care in the case of suprapubic bladder drainage
e) Tracheostomy care
Every child nursing doll is provided with a serial number. This is to be found on the inside in the centre of the chest. It is essential to quote this number in full when reordering.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty