Would you like to log in again?
Liquid Antiserums
Supplier: Aldon
IS3152 IS3150
470233-600EA 43.5 USD InStock
470233-600 470233-570
Liquid Antiserums
Educational Materials Biology Educational Materials Blood Typing Learning Activities
Liquid antiserums are pooled and ready to use.
  • Anti-A and B Serums Identify ABO Blood Group
  • Anti-Rho (D) Identifies + or - Rh Factor
  • No Refrigeration Necessary
  • Pooled
  • Ready-To-Use
Anti-A and B serums identify ABO blood groups (A, B, AB, or O), and anti-AB provides a check on type O determinations. Anti-Rho (D) identifies + or - Rh factor.
Refrigeration is not necessary. The antiserums are contained in dropper bottles. They are for educational purposes only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty