Would you like to log in again?
3B Scientific® Young's Modulus Apparatus
Supplier: 3B Scientific
3B Scientific®
1018527 1018528
470233-674EA 1209.69 USD InStock
470233-674 470233-676
3B Scientific® Young's Modulus Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Measuring apparatus for investigating the elastic deformation of rods of flat geometry and for determining the modulus of elasticity.
  • Load characteristics of the strain gauge may be ignored
  • Measurements can be set up with the material samples supported on both sides or clamped at one end
Measuring apparatus for investigating the elastic deformation of rods of flat geometry and for determining the modulus of elasticity. Features a strain gauge unit for determining the deformation of electrically conducting material samples. The strain gauge unit is electrically connected to the material sample in such a way that placing of the probes on the sample is determined with a high degree of sensitivity and displayed with the help of two LEDs. The bending of the material sample with the weight suspended is measured to an accuracy of 0.01 mm and the modulus of elasticity can then be calculated from the reading.
Experiment Topics – Elastic deformation of flat bars; Determining modulus of elasticity.
Advantages – Load characteristics of the strain gauge may be ignored; Measurements can be set up with the material samples supported on both sides or clamped at one end.
Delivery information: Contents includes 6 Flat steel rods (W: 15 mm, L: 200 / 300 / 400 mm, thickness: 2 / 3 mm); 1 Strain gauge unit; 1 Horizontal beam with stand; 2 Knife-edge bearings; 1 Clamping chuck; 1 Set of weights and retaining clamps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty