Would you like to log in again?
3B Scientific® Microwave Set 10.5 Ghz
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1009950
470233-704EA 2597.89 USD InStock
470233-704
3B Scientific® Microwave Set 10.5 Ghz
Educational Materials Physics Educational Materials Optics Learning Activities
Equipment set for conducting wave optics experiments involving wavelengths in the cm range.
  • Oscillator frequency: 10.5 GHz (U8493600-115)
  • Acoustic signal: Switchable
  • Receiver with horn antenna; Silicon diode with resonator
  • Microwave sensor; Silicon diode with resonator
  • Modulation type: 100 Hz to 20 kHz, 1 V max. (External modulation)
Equipment set for conducting wave optics experiments involving wavelengths in the cm range. A transmitter with a horn antenna radiates a narrow beam of linearly polarized electromagnetic waves with a wavelength of about 3 cm. The direction of polarization can be altered by rotating the antenna around the axis of propagation. To detect the waves, a horn antenna receiver and a microwave sensor are provided. A control unit converts the intensity of the signal received into a proportionally large output voltage that can be measured using a voltmeter. It is also possible to switch on an acoustic signal with a volume that is proportional to the intensity of the signal.
Oscillator frequency: 10.5 GHz.
Experiment topics – Straight-line propagation of microwaves; Reflection, absorption and transmission; Shielding from microwaves; Experiments on polarisation; Experiments on diffraction; Experiments on refraction and interference; Transmission of information.
Delivery information: Contents – 1 Control unit; 1 Transmitter with horn antenna; 1 Receiver with horn antenna; 1 Microwave probe; 1 Mikrowave bench, 800 mm; 1 Microwave bench, 400 mm with plate holder; 1 Reflection plate 180×180 mm²; 1 Polarisation grating, 180×180 mm²; 1 Absorption plate, fibreboard, 180×180 mm²; 1 Paraffin prism; 1 Stand for prism; 1 Plate with double slit; 1 Cover plate for double slit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty