Would you like to log in again?
Laser Diode for Debye-Sears Effect, 3B Scientific
Supplier: 3B Scientific
3B Scientific®
1002577 1002579
470233-726EA 718.09 USD InStock
470233-726 470233-728
Laser Diode for Debye-Sears Effect, 3B Scientific
Educational Materials Physics Educational Materials Optics Learning Activities
Laser diode of protection classification II with 1 m connector lead and barrel connector for connecting ultrasonic cw generator.
  • All lasers have been measured to determine their wavelength specifically and the results are logged
Laser diode of protection classification II with 1 m connector lead and barrel connector for connecting ultrasonic cw generator (U100061). Fits the laser holder of test vessel (U10008). All lasers have been measured to determine their wavelength specifically and the results are logged.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty