Would you like to log in again?
Induction Tube Apparatus with 6 Coils, 3B Scientific
Supplier: AMERICAN 3B SCIENTIFIC SE
1001005
470233-824EA 301.79 USD InStock
470233-824
Induction Tube Apparatus with 6 Coils, 3B Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Guinea and feather tube apparatus with 6 induction coils connected in series.
  • Guinea and feather tube apparatus with 6 induction coils connected in series
  • The voltage characteristic can be plotted using any conventional storage oscilloscope or with a computer interface
Guinea and feather tube apparatus with 6 induction coils connected in series. When a bar magnet (included in the scope of delivery) is dropped in the tube, a voltage is induced as it passes each of the respective coils. Due to the magnet's free-fall acceleration caused by gravity, the voltage peaks increase proportionally with the time (speed proportional to time), and the time intervals between the voltage peaks become ever shorter. The voltage characteristic can be plotted using any conventional storage oscilloscope or with a computer interface.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty