Would you like to log in again?
Coulombmeter with Rechargeable Battery, 3B Scientific
Supplier: 3B Scientific
3B Scientific®
1017734
470233-864EA 391.89 USD InStock
470233-864
Coulombmeter with Rechargeable Battery, 3B Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Coulombmeter for measuring the flow of current when storing energy in a rechargeable battery.
  • Charge or consumption can be displayed by means of a voltmeter
  • 4 mm Safety Sockets
Depending on the measuring range selected, 1 V on the voltmeter corresponds to either 0.1, 1 or 10 A/s.
Load current: max. 500 mA; charging current for battery: max. 50 mA
Power supply: 9 V rechargeable battery via DC co-axial power socket; External power source: Solar panel or DC power supply (max. 12 V DC) with current limiting to 50 mA when there is no load on the coulombmeter
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty