Would you like to log in again?
BristleBot Kit
Supplier: BROWN DOG GADGETS
K1003-25PK K1003-10PK K1003-4PK
470234-642PK 19.95 USD InStock
470234-642 470234-644 470234-646
BristleBot Kit
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Teach STEAM with ease!
  • Build time 5 to15 minutes
  • Age range 9+
  • Tools needed: none
Bristlebots are small robots made by combining a toothbrush head and vibrating motor. By changing the position of the pipe cleaner arms, Bristlebots will travel in circles, straight lines, or scoot around randomly. What can you make your Bristlebot do? Bristlebots are great for teaching engineering, motors, circuits, and principles of balance.
Ordering information: Bristlebots do not require tools to build. The toothbrush head comes pre-cut, and motor wires have exposed ends to connect to the battery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty