Would you like to log in again?
Solar USB Kit 2.0
Supplier: BROWN DOG GADGETS
K1011
470234-658EA 47.35 USD InStock
470234-658
Solar USB Kit 2.0
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Teach any child or student the power of solar energy by building a USB solar charger!
  • Build time 30 to 60 minutes
  • Suitable for grades 8+
  • Requires basic soldering knowledge
The finished charger is capable of charging small electronic devices. Three rechargeable AA batteries, that when soldered to the solar panel, create a charging circuit that allows solar energy to be stored in the batteries. A Baltic Birch enclosure houses all of the internal parts and is held together by screws. We recommend painting or staining your enclosure to add a personal touch to it.
Note: Additional USB Cable not needed. It's an option in case you need one. When in doubt use the USB cable that came with your gadget.
Ordering information: Parts include USB Charging Circuit - works with all iPhone and iPod models, as well as most other phones and USB gadgets; 6V 80 mA Solar Cell; 3 AA Holder; 3 Rechargeable AA Batteries @ 2600 mAh; Toggle Switch; Wire; and Laser Cut Enclosure with Screws.
Items Needed: Soldering Iron; Double-sided Foam Mounting Tape; Screwdriver; Wire Stripper or Cutter; and Hot Glue Gun.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty