Would you like to log in again?
biOrb® Air Stone
Supplier: GEOGLOBAL PARTNERS
46029
470234-876CS 59.4 USD InStock
470234-876
biOrb® Air Stone
Animal Supplies Animal Environmental Enrichment
The biOrb® Air Stone fits in the base of the biOrb® underneath the filter cartridge.
  • Breaks up the flow of air into the aquarium, splitting it into a mass of fine bubbles
  • Reduces noise from the air pump
Over time the tiny pores in the air stone can become blocked by dirt and lime scale preventing air from getting through properly. Depending on your local water supply and the conditions in your biOrb® an air stone can last anywhere from 3 months to 3 years. Changing the air stone from time to time is advisable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty