Would you like to log in again?
biOrb® Aquarium Sculptures
Supplier: GEOGLOBAL PARTNERS
46122 46128 46123 46124 46126 46121 46141 46131 46107 46140 46108 46127 46116 46132 46145 46130 46146 46103 46109
470235-006CS 91.85 USD InStock
470235-006 470235-017 470235-051 470235-041 470235-063 470235-061 470235-091 470235-081 470235-015 470235-059 470235-013 470235-079 470235-047 470235-055 470235-045 470235-089 470235-053 470235-031 470235-043
biOrb® Aquarium Sculptures
Animal Supplies Animal Environmental Enrichment
A centerpiece display for your aquarium.
  • Fit around the filter tube
  • Inert composition, safe for plants and animals
  • Use in marine or freshwater tanks
These sculptures allow you to add a beautiful centerpiece to your biOrb® aquarium. Each sculpture is designed to fit around the central filter tube without affecting the filtration system. Each sculpture is made from an inert resin and will retain its appearance for many years. If you don't have a biOrb® aquarium, that's ok. The central opening provides a great hiding place for small fish or shrimp.
Available in four sizes, the small sizes are lower and suitable for any biOrb® from 15 l up. The Medium fit a 30 l or larger biOrbs. Large should be used with the Classic and Life series biOrbs of 45 l or larger. The Extra Large are designed to fit the additional height of the 60 l and larger. If you have a traditional aquarium, please consult the height information for a proper fit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty