Would you like to log in again?
biOrb® Air Terrarium
Supplier: GEOGLOBAL PARTNERS
46147 46148
470235-094EA 541.65 USD InStock
470235-094 470235-092
biOrb® Air Terrarium
Animal Supplies Animal Environmental Enrichment
A Compact, Easy To Manage Terrarium.
  • 360 degree view
  • Automatic misting
  • LED lighting
The biOrb® AIR Terrarium is THE modern terrarium/vivarium, whether it's orchids, poison dart frogs, air plants, mosses and carnivorous plants or an insectarium the biOrb® AIR replicates the conditions found under the tropical forest canopy and will provide them with the humidity, air circulation and lighting they need. You also get a 360 degree view of a world you've created and the pleasure of watching it develop and grow, it's like a window into another world. The biOrb® AIR uses treated sterilized coir compost as the growth media to cultivate the plant roots. The coir compost is placed on top of a capillary mat that draws water up to the compost from a small water reservoir in the base. The capillary action prevents the plant roots from soaking and rotting. Natural daylight is not necessary as the lighting is arranged in a six LED array at the top of the sphere. This emits the perfect light output for encouraging natural growth and prevents plants bending towards a single light source. The light also works on an automatic 24 hour cycle, simulating sunrise, daylight, sunset and night time. To discourage stagnation and condensation within the biOrb® AIR, a constantly running fan, with three speed options: low, medium and high, recycles the air through a replaceable carbon filter. A small amount of fresh air is regularly introduced as part of the circulation process. To maintain the humidity within the micro-climate, water is added using an ultrasonic mister mounted in the top water reservoir. This mister can be set to three different settings: automatic for tropical plant growth, low for a less humid environment and high for certain amphibian and invertebrate animal species. The terrarium is available in glossy white or grey.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty