Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
OHAUS® Light Duty Orbital Shakers
Supplier: Ohaus
Ohaus
30400119 30400099 30400185 30400100 30400190 30400157 30400182 30400152 30400116 30391924 30391931 30400088 30400151 30400189 30400191 30400188 30400187 30400183 30400156 30400086 30400115 30391945 30391938 30400193 30400084 30400184 30400159 30400114 30400085 30400158 30400194 30400120 30400168 30400186 30400087 30400172
470235-208EA 191 USD InStock
470235-208 470235-228 470235-206 470235-226 470235-204 470235-162 470235-160 470235-224 470235-202 470235-324 470235-222 470235-200 470235-288 470235-322 470235-300 470235-220 470235-342 470235-340 470235-218 470235-216 470235-294 470235-292 470235-334 470235-158 470235-236 470235-214 470235-156 470235-332 470235-212 470235-234 470235-198 470235-210 470235-232 470235-298 470235-296 470235-230
OHAUS® Light Duty Orbital Shakers
Shakers and Mixers Shaking Incubators
Save Incubator Space with Benchtop Orbital Shakers.
  • LED displays for temperature, speed and time
  • Timer with audible alarm
  • Calibration mode for temperature
  • Microprocessor-controlled for consistent shaking action
  • Triple eccentric drive provides reliable service and continuous duty
  • Safety features include speed ramping and overload protection
Ohaus Incubating Light Duty Shakers are designed to incubate samples from 10° below ambient to 65 °C depending on the model. The incubating Mini shaker has a 3.6 kg (8 lb.) capacity while the Incubating Microplate Shaker can hold up to four standard or deep well plates. Incubating-Cooling Mini Shaker is designed to hold two microplates or two optional modular tube blocks. All models feature LED displays with touchpad controls.
Operating Features: Microprocessor Control-The variable speed microprocessor control provides consistent uniform shaking action. Microprocessor will display last set-point and will restart if power is interrupted. PID Temperature Controller–Maintains precise temperature control from ambient +5 to 65 °C. Easy-to-use controls allow users to adjust temperature in 1 °C increments. Triple Eccentric Drive-Permanently lubricated ball bearings and maintenance free, brushless DC motor provide reliable service and continuous duty operation. LED Display-Touch pad controls with easy-to-read, independent LED displays for temperature, speed and time allow operator to view all settings at once. Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off. Temperature Calibration Mode-Allows user to calibrate unit to an external temperature device.
Safety Features: Overload Protection-Audible and visual signals will activate when system detects an obstruction or overload of the tray. Speed Ramping Feature–Slowly increases speed to desired set-point to avoid splashing. Audible Alarm-In timed mode, alarm will sound when the time reaches zero. Caution Hot Indicator-Symbol illuminates when the temperature of the air in the chamber reaches 40 °C and remains lit until temperature cools down. Spill-Resistant Design-Channels fluids away from internal components. Polycarbonate Lid-Permits viewing of samples without disturbing internal temperature.
Operating Conditions–Unit can be run in conditions from 5 to 40 °C (41 to 104 °F), maximum 80% relative humidity, non-condensing. Applications–Cell cultures, bacterial suspensions, hybridizations, Immunoassays.
Ordering information: Unit includes a detachable, 3-wire cord and plug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty