Would you like to log in again?
OHAUS® Heavy Duty Vortex Mixer
Supplier: Ohaus
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ohaus
30400226 30400214 30400211 30400210 30400218 30400220 30392146 30400209 30400224 30400216 30400215 30400222 30400221 30400212 30400223 30400213 30400217
470235-370EA 59 USD InStock
470235-370 470235-392 470235-382 470235-390 470235-380 470235-368 470235-376 470235-388 470235-366 470235-374 470235-364 470235-166 470235-386 470235-394 470235-372 470235-384 470235-362
OHAUS® Heavy Duty Vortex Mixer
Shakers and Mixers Vortex Mixers
Designed for Continuous Duty Applications.
  • LED displays for speed and time
The OHAUS Digital Heavy-Duty Vortex Mixer is ideal for applications that demand repeatable results. Mixer features touchpad controls and LED displays for accurate speed (rpm) and time (minutes and seconds) results. The heavy-duty design and efficient motor allow this mixer to operate in continuous duty and handle all accessories over the entire speed range. Choose from two modes of operation- "Touch" mode for mixing tubes when cup head or Universal Holder with cover is depressed, or "On" mode when using any of the accessory attachments for continuous operation. Microprocessor control maintains set speed for strong, consistent mixing action. Timer will display elapsed time or, when programmed to user defined time limits, the unit will shut off when time reaches zero.
Operating Features: Microprocessor Control-The variable speed microprocessor control provides consistent uniform mixing action. LED Display-Touch pad controls with easy-to read, independent LED displays for speed and time allow operator to view both settings at once. Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.
Operating Conditions: Unit can be run in conditions from 4 to 40 °C (39 to 104 °F), 20 to 85% relative humidity, non-condensing.
Ordering information: Mixer includes a cup head, universal holder with cover, foam microtube insert for (38) 1.5 to 2.0 mL microtubes, and a detachable, 3-wire cord and specified plug.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty