Would you like to log in again?
Demonstration Pulleys
Supplier: G.S.C. INTERNATIONAL, INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4218-00-10 4217-00-10 4219-00-10
470235-700EA 9.05 USD InStock
470235-700 470235-698 470235-696
Demonstration Pulleys
Educational Materials Physics Educational Materials Mechanics Learning Activities
Jumbo-Sized Pulleys are Designed for Classroom Demonstrations
These pulleys are open to make it easier for users to run pulley cords through them. The single, double, and triple pulleys are all 2 3/4" in diameter. Light-weight, low-friction, and deformation-resistant plastic ensures accurate results.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty