Would you like to log in again?
Cartesian Diver
Supplier: GSC INTERNATIONAL SE
305-10
470235-702EA 12.35 USD InStock
470235-702
Cartesian Diver
Educational Materials Chemistry Educational Materials Gas Analysis Learning Activities
Ideal for Showing Archimedes’ Principle
Students will be fascinated at their first encounter with our Cartesian Diver. We supply a hollow glass diver, even shaped like a diver, with a 10" x 1 1/2" glass cylinder, a rubber sheet and rubber band. Fill the cylinder with water, drop in the diver so he floats, fasten the sheet tautly across the top and then gently depress the sheet. The diver will dive, his speed depending upon the pressure you place on the sheet; release the sheet and the diver returns to the surface. The diver’s behavior illustrates the uniform transmission of pressure in all directions, the compressibility of gases and Archimedes’ Principle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty