Would you like to log in again?
Eudiometer
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG2202/1 503-2-10
470235-712CS 577.1 USD InStock
470235-712 470219-674
Eudiometer
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Demonstrate the combining volume ratios of gases.
  • Two sizes available
  • Platinum electrodes included
Long, sturdy glass tubes have platinum electrodes sealed at the closed end with a 2 mm arc gap. The tubes are marked with a permanent brown scale in 0.1 or 0.2 mL divisions for 50 and 100 mL respectively. An induction coil is recommended for use with eudiometers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty