Would you like to log in again?
Borosilicate Glass Geissler Burets
Supplier: GSC INTERNATIONAL SE
9380-2-5 BR2124-50 BR2124-50 BR2124-100 9380-3-5 BR2124-25
470235-716EA 60.2 USD InStock
470235-716 470235-714 470201-420 470148-958 470104-058 470104-056
Borosilicate Glass Geissler Burets
Burets
Student grade buret with fitted glass stopcock.
  • Glass tubing is annealed to relieve stress
  • Divisions are clearly marked
Burettes are made from the highest quality accurate bore tubing to provide a high degree of accuracy, reliability, and durability. Amber graduations. Includes a straight bore glass stopcock. Accuracy as per Class B.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty