Would you like to log in again?
Electrolysis Electrodes
Supplier: G.S.C. INTERNATIONAL, INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
501-2-10
470235-734EA 77.55 USD InStock
470235-734
Electrolysis Electrodes
Electrodes Other Electrodes
Platinum sealed in glass tubes inserted securely in a rubber stopper. Electrodes fit standard electrolysis tubes. It has a rubber stopper No.1 with 18 gauge copper wire.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty