Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quality Corks, XX

Supplier: G.S.C. INTERNATIONAL, INC.

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CS-6-100-10 CS-4-100-10 CS-2-100-10 CS-3-100-10 CS-20-10
470235-706EA 7.4 USD InStock
470235-706 470235-704 470235-732 470235-744 470235-742
Quality Corks, XX
Stoppers Cork Stoppers
Size Top Diameter Bottom Diameter

Size 1 11.1 mm 8.3 mm

Size 2 12.7 mm 9.5 mm

Size 3 14.2 mm 10.7 mm

Size 4 15.8 mm 11.9 mm

Size 6 19.0 mm 14.6 mm

Size 20 41.2 mm 34.0 mm

  • Size 2 - sold in a case; 10 packs of 100
  • Size 3 - sold in a case; 10 packs of 100
  • Size 4 - sold in a case; 10 packs of 100
  • Size 20 - sold in a pack of 10
  • The natural cork is safe for use with food products

Get the perfect fit with our lab-grade cork stoppers. These stoppers ensure a secure, leak-proof seal every time. Crafted from natural cork, these stoppers are safe for use with food products, making them a reliable choice for all your storage needs.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty