Would you like to log in again?
Flint Glass Y-Shaped Connecting Tubes
Supplier: GSC INTERNATIONAL SE
2004-4-10
470235-774EA 6.6 USD InStock
470235-774
Flint Glass Y-Shaped Connecting Tubes
Fittings and Connectors
Y-shaped connector with a 120° angle provides safer connections as the ends have constrictions to hold rubber tubing securely.
  • Made from quality flint glass
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty