Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Sterile Sampling Bags
Supplier: Ward's
Ward's®
B5761_WARDS B5780_WARDS B5790_WARDS B5771_WARDS B5750_WARDS B5911_WARDS
470236-306PK 198.9 USD InStock
470236-306 470236-308 470236-302 470236-304 470236-298 470236-300
Ward's® Sterile Sampling Bags
Bags Sample Bags
Gamma sterilized bags make sample collection quick and easy.
  • Each sterile bag is individually sealed with a tear-off closure
  • Extended PVC tab covers the stainless steel wire, to eliminate glove punctures
  • Depending on the size, these bags can hold up to 1600 ml (55 oz.)
Samples can be easily collected by removing the tear strip above the wire closure and pulling on the two external grip tabs to open the mouth of the bag to the desired level. Using the grip tabs ensures sterility and eliminates the risk of contamination from finger contact. Then, introduce the sample into the bag, flip or fold the bag three times, bend the wires and the sample is secure inside the airtight and moisture sealed bag.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty