Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mobile Draw Station

Supplier: MARKETLAB, INC.

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

42150 10016
470236-370EA 185.09 USD InStock
470236-370 470236-368
Mobile Draw Station
Trays/Dishes/Pots Blood Collection Trays
Create a draw station anywhere.

  • Increase efficiency by keeping supplies in full view and as close to the patient as possible
  • Great organization solution for any small area
  • Heavy-duty spring securely clamps on chair arms, wheelchairs, and bed rails

If you have been a phlebotomist for very long, you know many patients are physically unable to make it to a draw room. As a result, these patients must be drawn in alternate locations that are often awkward and unsafe. The Mobile Draw Station was designed by a group of phlebotomists who wanted a portable "workstation" that could travel anywhere.

Delivery information: Mobile Draw Station Plus included 6 x 17 mm tubes, 4 x 14 mm tubes, 2 x 11 mm tubes, alcohol wipes and adhesive bandages.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty